3M™ DF Series Filter Cartridge
Supplier: 3M Healthcare
7100018269 7000029181 7000050782 7000029255 7000050483 7010338872 DFG005FE2C 7000001607 7100010838 7000029183 7000029184 7000050485 7000029256 7000050757 7010380927 7000050239 7000050240 7000029185 7000001608 7000050486 7000029257 7000050487 7000050783 7000050241 7000050243 7100011131 7000001698 7000050785 7000050478 7000029252 7000050479 DFG001FE2C DFG001FE2R 7000050482 7000050237 7000029254 7000050242 7000029186 7000001697 7000050489 7000050490 7100011134 7000050491 7100010306 7000050492 7000050493 7000050494 7010314630 7100010305 7000050245 7100011097 7000001609 7000050786 7000125619 7000050495 7000050787
76029-106CS 564.52 USD
76029-106 76030-333 76030-335 76030-337 76030-339 76030-341 76030-343 76030-347 76030-349 76030-351 76030-353 76030-355 76030-357 76030-359 76030-361 76030-363 76030-365 76030-367 76030-369 76030-371 76030-373 76030-375 76030-377 76030-379 76030-381 76030-383 76030-385 76030-387 76032-877 76032-879 76032-881 76032-883 76032-885 76032-887 76032-889 76032-891 76032-893 76032-895 76032-897 76032-899 76032-901 76032-903 76032-905 76032-907 76032-909 76032-911 76032-913 76032-915 76032-917 76032-919 76032-921 76032-923 76032-925 76032-927 76032-929 76032-931
3M™ DF Series Filter Cartridge
Filters Filter Cartridges
3M™ DF series filter element is a proven alternative to most conventional bag filters. Our filter is a true graded porosity media structure that provides a 62% increase in surface area compared to a standard #2 bag and up to four times more service life.
  • Graded–porosity filter medium provides up to four times more life than most conventional bag filters for fewer change–outs
  • Easily retrofits standard bag filter housings
  • Reduced filter changes help minimize product loss, labor, disposal costs, and operator exposure.
  • Cartridge design reduces filter medium rupture, contaminant by–pass and unloading
  • Simplifies filter installation, removal and disposal
The 3M™ DF series filter media consists of 2 layers with the first layer being 'open' to remove the larger contaminants while the second layer is 'tighter' to remove the smaller contaminants
The 3M DF series filter element consists of two cylinders bonded to a top plate and a lower seal plate. As fluid enters the top of the filter through flow channels in the top plate, it flows between the inner and outer media cylinders. Then it passes through the media and support basket and into the clean chamber of the filter housing. Our design incorporates an innovative geometry of both bag filter and restrainer basket, giving complete three–dimensional support of the media. This eliminates the potential for bag filter rupture and the resulting gross contamination of the downstream effluent. The 3M DF series filter element design reduces hold–up fluid volume by 67% compared to conventional filter bags, which helps minimize worker exposure to process fluids due to less frequent bag change–outs.
3M™ DF Series Filter Best Features: 100% downstream support of the filter element, eliminates filter rupture, contaminant bypass and unloading, allows operation to higher differential pressures before filter change–out, Superior flow characteristics, maximizes utilization of filter surface area and maintains low operating pressure drop, Reduces flow per unit area (flux) for improved effluent quality, Greater media surface area, Achieves up to four times the service life as compared to conventional filter bags, Improves the retention efficiency of the bag filter, Greater contaminant holding capacity, Media layers of different porosities are combined to form a graded porosity structure, Each nominally rated filter medium has been optimized to achieve the longest service life, We use advanced thermal sealing processes to weld the side seams to form the bag and secure the sealing ring. Thermal–singe treatment of the exterior surface prevents material fibers from migrating into the clean filtered process media. Each grade of the 3M™ DF series filter element is manufactured from high performance fibers that are selected, based on extensive media performance testing. No adhesives, binders or silicone are used in the manufacturing processes.
Applications include: Industrial waters, Lubricants, Chemicals, Petrochemicals, Food, Wine filtration, Beer micro filtration, Process Water, Pharmaceuticals, Electronics, Purification, Water Treatment.