3M™ Betapure™ BK-Z8 Series Filter Cartridge
Supplier: 3M Healthcare
BK09Z8100G1SNG BK30Z8100G1SNG BK40Z8050G1SFK BK19Z8050G1SNG BK19Z8070G1SNG BK19Z8100G1SNG BK20Z8050G1SNG BK20Z8100G1SNG BK29Z8100G1SNG
76029-733CS 600.1 USD
76029-733 76029-755 76029-775 76032-155 76032-157 76032-159 76032-163 76032-165 76032-181
3M™ Betapure™ BK-Z8 Series Filter Cartridge
Filters Filter Cartridges
Betapure™ BK–Z8 series filter cartridge is a truly absolute–rated, rigid, or non–compressible, resin–bonded filter cartridge. Rigid, resin–bonded structure prevents by–pass and unloading at high differential pressure. This product is available in lengths ranging from 9¾ to 40" in both standard and high temperature options.
  • Absolute rated filter cartridge, available in grades from 5 to 70 µm
  • Rigid, resin–bonded structure prevents by–pass or unloading at high differential pressure
  • Grooved surface with true graded–porosity internal structure provides long life
  • 300 °F high temperature option to meet the requirements of a wide–range of applications
An optimized groove pattern in Betapure BK–Z8 series filter cartridges to increase its surface area. The grooved surface prevents premature blinding of the outer surface by large particles and allows full utilization of the depth structure. Maximum surface area with a true graded–porosity structure means that Betapure BK–Z8 series filter cartridge can provide long service. Polypropylene or polyester end modifications can be retrofitted to any industrial housing and are usable in existing filter housings. The rigid design needs no plastic or metal core. The structure also contributes to consistent throughput quality throughout the filter's life. Our control process results in consistent filter cartridges with defined absolute ratings time after time after time. We offer the Beta–Klean Z8 series filter cartridge in a standard version that provides consistent performance to 250 °F (121 °C), as well as a high temperature option that extends performance to 300 °F (149 °C).
Applications: Betapure BK–Z8provides consistent reproducible filtration performance and long lifein a wide variety of industrial processing applications. Betapure BK–Z8 is particularly well suited for high viscosity fluids, high temperature processes and maximum differential pressures to 70 psid (4.8 bar) at (68 °F) 20 °C, Petroleum Products, Gasoline, Kerosene, Lube, Oil, Fuel oil, Wax, Chemical/petrochemical, Acids, Bases, Organic solvents, Catalysts, Monomers, Polymers, Glycols, Water, Process water, Produced water, Boiler feed, Demineralized feed, Pre–reverse osmosis system, Waste water, General industrial, Paint, Varnish, Lacquer, Inks, Coatings, Emulsions, Magnetic media, Resins, Detergents, Adhesives, Brines, Aqueous salt solutions.
Part of Betapure BK–Z8 series filter cartridge's efficiency is due to its rigid, resin–bonded structure, which helps retain contaminants in dynamic processes, such as high differential pressure, with no by–pass or unloading. We construct each graded–porosity filter cartridge using acrylic fibers, cellulose fibers and a tough chemically resistant thermosetting resin. Our manufacturing process produces more fibers towards the center core region, creating a graded–porosity structure, and the thermosetting resin "bonds" the fibers into a permanent, rigid matrix.