3M™ Zeta Plus™ BC Series Filter Capsule with Activated Carbon Series Media
Supplier: 3M Healthcare
BC0025LR52SP
76029-582CS 308.45 USD
76029-582 76029-586 76029-592 76029-594 76029-596 76029-706 76029-573 76029-575 76029-577 76029-579 76029-581 76029-583 76029-585 76029-587 76029-589 76029-591 76029-593 76029-595 76029-597 76029-599 76029-601 76029-653 76029-655 76029-657 76029-661 76029-663 76029-665 76029-667 76029-669 76029-671 76029-673 76029-675 76029-677 76029-679 76029-681 76029-683 76029-685
3M™ Zeta Plus™ BC Series Filter Capsule with Activated Carbon Series Media
Filters Capsule Filters
Zeta Plus™ BC Series Filter Capsule with Activated Carbon Series Media contains activated carbon to decolorize and remove contaminants from process streams. The media can be used in applications where bulk activated carbon is used. Activated carbon particles and cellulose fibers mechanically filter contaminants while a positively charged polymer resin may adsorb negatively charged particles.
  • Totally enclosed, sanitary design filter capsule significantly reduces time required for cleaning
  • In addition to activated carbon adsorption the filter media also may retain contaminants by mechanical entrapment and electrokinetic adsorption
  • Single-use capsule reduces the hassle of cartridge change outs and eliminates housing cleaning validation
  • All product components comply with United States Pharmacopeia (USP) Class VI 70 °C Biological Reactivity, in vivo
We offer five standard types of activated carbon to cover a broad range of applications.
Our activated carbon media is also offered in three porosities to enable processing of solutions with different viscosities.
Scalability.
Three capsule sizes are offered having surface areas of 25, 650 and 1300 cm². The effective lenticular filter and vertical flow path design is preserved on these two larger Zeta Plus™ BC series filter capsules. The designs have proven effective for depth filtration systems and for consistent performance for all sizes.