3M™ LifeASSURE™ PLA Series Filter Capsule
Supplier: 3M Healthcare
7000050335 7100139039 7000050331 7000050332 7100109454 7010337850 7010296351 7010315116 7010380685 7000001635 7000029216 7000050336 7000050337 7000050965 7000050338 7010338512 7010290696 7010380955 7100122478 7000050340 7010415182 7010380690 7010381296 7000029355 7010314298
76032-754CS 4559.25 USD
76032-754 76034-100 76034-102 76034-104 76034-106 76034-108 76034-110 76034-112 76034-114 76034-116 76034-118 76034-120 76034-122 76034-124 76034-126 76034-132 76034-134 76034-136 76034-138 76034-140 76034-148 76034-150 76034-152 76034-154 76034-156 76034-158 76034-160
3M™ LifeASSURE™ PLA Series Filter Capsule
Filters Capsule Filters
LifeASSURE™ PLA Series Filter Capsules encompass two important design elements-multi‐zone microporous membrane and Advanced Pleat Technology-to offer prefiltration protection of final membrane filters. These filters have applications in prefiltration and clarification in the pharmaceutical, biological and bioprocess industries.
  • Multi-zone microporous nylon membrane offers maximum membrane protection and increased contaminant capacity
  • Advanced Pleat Technology construction allows maximum fluid and contaminant access to filter surface area for highest throughput
  • 100% Integrity tested prior to release ensures consistent performance to specification
Multi-zone microporus membrane construction.
We incorporate our multi‐zone microporous membrane technology into our LifeASSURE™ PLA Series filter capsules. This technology allows great flexibility in creating a multi‐zone membrane that offers excellent contaminant holding capacity while maintaining high retention efficiency. LifeASSURE™ PLA series filter membrane consists of an 'open' zone on the upstream side of the membrane and a 'tighter' zone on the downstream side. In effect, the open zone acts as a prefilter by capturing larger particles and colloids.
Advanced pleat technology design.
LifeASSURE™ PLA Series filter capsules also feature our Advanced Pleat Technology (APT) design for extended service life. This design technology is intended to increase the useful surface area while maintaining open flow paths between media pleats.
We manufacture LifeASSURE™ PLA Series filter capsules using all polypropylene structural components (end caps, adapters, media support, inner core and outer cage). The 3M Purification Inc. Quality Management System is approved by an accredited registering body to the ISO 9001:2008 and ISO 13485:2003 Quality Management Systems, and the cartridges are 100% forward flow integrity tested prior to shipment. All LifeASSURE PLA Series filters are shipped with a Certificate of Quality affirming compliance with manufacturing quality specifications. Supporting Drug Master File (DMF) documentation is on file with the United States Food and Drug Administration (FDA).