3M™ Zeta Plus™ BC Series Filter Capsule with S Series Media
Supplier: 3M Healthcare
7010338579 7010413201
76029-621CS 750.39 USD
76029-621 76032-133
3M™ Zeta Plus™ BC Series Filter Capsule with S Series Media
Filters Capsule Filters
Full range of scaleable capsule and cartridge filter configurations allow pilot testing and scale-up with the same materials that will be used in full-scale systems and electrokinetic adsorption. Full range of scaleable capsule and cartridge filter configurations allow pilot testing and scale-up with the same materials that will be used in full-scale systems.
  • High contaminant holding capacity for economical filtration and reliable particle reduction
  • Retains contaminants by mechanical entrapment and electrokinetic adsorption
Self-contained, single-use disposable capsules and cartridge modules reduces labor time for change-outs and eliminates cleaning validation.
Ideal for adsorptive depth filtration, beer clarification, beer microfiltration, clarification of concentrates, ingredients and beverages, Haze reduction for distilled spirits, keg filtration, microbial reduction filtration, particle reduction in distilled spirits filtration, wine clarification.