3M™ CT Series Filter Housing
Supplier: 3M Healthcare
7000029344 7000125372 7100066076
76031-816CS 2132.84 USD
76031-816 76031-818 76031-820
3M™ CT Series Filter Housing
Filters Filter Cartridges
3M™ CT series cartridge filter housings are three-piece, all-metal housings with ring nut closures, designed to securely hold a range of filter types to meet your industrial and process filtration needs. Available in both 304 stainless steel and 316 stainless steel material construction to meet various application needs.
  • Rugged, three-piece, all metal construction for durability
  • Efficient design provides quick and easy cartridge change-out
  • Design flexibility allows use of 1, 2 or 3-high cartridge stack
  • Drilled and tapped mounting holes are ready for
More Product Information