3M™ DC Series Filter Housing
Supplier: 3M Healthcare
7000125748 7000125749 7100066574 7000125750 7000029483 7000125751 7000125752 7010339748 7100031449 7010382829 7010340337
76029-016CS 1957.76 USD
76029-016 76029-018 76029-020 76029-022 76029-024 76029-026 76029-028 76029-038 76029-040 76029-048 76029-050
3M™ DC Series Filter Housing
Filters Filter Cartridges
3M™ DC series filter housings, constructed of durable 304L stainless steel, meet general–purpose industrial and commercial filtration needs.
  • 304L stainless steel construction provides excellent corrosion protection for rugged, long–lasting service
  • Allows use of 9¾ to 10" modular cartridges for greater flexibility
  • Solid fit into cartridge core for a positive seal and helps prevent bypass
  • V–Band Clamp and Swing bolt designs optimize cartridge change–out
  • Allows full drainage of chambers for optimized filter change–out
  • Air Vent allows venting before start–up and draining for more efficient filter operation
  • Allows for use of both DOE or SOE cartridges
The filter housings are available in a variety of sizes to accommodate from 4 to 88 filter cartridges (Single length equivalents) with lengths to 40". The 4DC and 5DC housings handle liquids at pressures and temperatures of up to 150 psig (10.3 bar) at 121 °C (250 °F) and the 12DC and 22DC housings handle liquids at pressures and temperatures of up to 150 psig (10.3 bar) at 212 °C (100 °C).
All wetted welded surfaces are stainless steel. External surfaces of these filter housings are electro–polished for a consistent, easy care finish.