3M™ Betafine™ DP Series Filter Cartridge
Supplier: 3M Healthcare
7010409162 7010410991 7010380997 7010380383 7010339561 7000050248 7010409033 7010413186 7000050247 7010379431 7010379433 7010380279 7010381356 7010338967 7010338034 7010379436 7010339317 7010314646 7010409447 7010381704 7010338756 7010379441 7010413238 7010338038 7010314870 7010338771 7010413600 7010339362 7010381326 7010339920 7010338636 7100083580 7010338706 7010411116 7000125739 7010301310 7000050526 7010379447 7100111883 7000050250 7010383144 7010301308 7010339715 7000050820 7100008197 7010379865 7010313731 7000050528 7010383128 7010412760 7010413004 7100111885 7010379860
76028-567CS 2398.07 USD
76028-567 76028-569 76030-389 76030-395 76030-397 76030-399 76031-569 76031-571 76032-933 76032-935 76032-943 76032-945 76032-953 76032-959 76032-961 76032-963 76032-967 76032-971 76032-979 76032-981 76032-995 76032-999 76033-001 76033-003 76033-005 76033-007 76033-009 76033-011 76033-015 76033-019 76033-021 76033-027 76033-029 76033-031 76033-033 76033-035 76033-043 76033-045 76033-047 76033-051 76033-053 76033-055 76033-057 76033-063 76033-067 76033-069 76033-075 76033-077 76033-079 76033-649 76033-651 76033-653 76033-655
3M™ Betafine™ DP Series Filter Cartridge
Filters Filter Cartridges
Betafine™ DP series pleated polypropylene filters are available with absolute ratings from 0.2 to 70 µm. The all–polypropylene filter is used extensively on corrosive and non–corrosive process fluids, where broad chemical compatibility is required.
  • Absolute–rated pleated filter provides consistent, repeatable filtration performance
  • Pleated media offers greater surface area
  • 100% polypropylene construction for broad chemical and thermal compatibility
  • FDA, CFR21 listed materials of construction, suitable for food, beverage, and pharmaceutical applications
More Product Information