3M™ Betafine™ XL Series Filter Cartridge
Supplier: 3M Healthcare
7010415285 7010409849 7010410984 7010413407 7010412829 7010410985 7000050543 7000050570 7000148454 7000051020 7010381701 7000050586 7100008814 7010339082 7100006943 7000050802 7000029302 7010380242 7000050589 7000050590 7000050803 7000050866 7000125733 7010380412 7010339086 7010380426 7000050544 7000050763 7000050546 7000050547 7000051251 7010339378 7010415233 7000029292 7100019936 7000050764 7100030799 7100060349 7000050562 7000051131 7010338188 7100026655 7100006939 7000050846 7000051710 7010296370 7000050768 7010379991 7100007520 7000050575 7000029296 7010338195 7100049398 7010314810 7010413562 7010381515 7000125714 7100021775 7100095539 7000050581 7010339710 7000125715
76028-634CS 1875.95 USD
76028-634 76028-631 76028-633 76031-613 76031-617 76031-619 76033-147 76033-341 76033-343 76033-347 76033-477 76033-479 76033-481 76033-483 76033-485 76033-487 76033-489 76033-491 76033-495 76033-499 76033-501 76033-505 76033-509 76033-513 76033-515 76033-517 76033-519 76034-419 76034-421 76034-423 76034-427 76034-433 76034-435 76034-439 76034-441 76034-443 76034-445 76034-447 76034-449 76034-451 76034-453 76034-457 76034-459 76034-461 76034-467 76034-471 76034-473 76034-475 76034-477 76034-479 76034-481 76034-483 76034-485 76034-487 76034-489 76034-491 76034-493 76034-495 76034-497 76034-499 76034-501 76034-503 76034-505
3M™ Betafine™ XL Series Filter Cartridge
Filters Filter Cartridges
Betafine™ XL Filter Cartridge Series is made of a 100% polypropylene pleated filter featuring Advanced Pleat Technology (APT). This product is appropriate for use in a wide range of applications, including: food and beverage, industrial and electronics.
  • Available in a range of integral lengths from ⁹/₃₄ to 40 inches, with a wide selection of end treatments to fit common filter housing designs
  • 100% polypropylene materials of construction for chemical and thermal compatibility
  • Absolute–rated cartridge filter available in nine distinct filter grades
Betafine™ XL series filter cartridge features 100% polypropylene construction that provides chemical and thermal compatibility in many demanding process fluid applications.
Pleated filters may offer a large gross surface area, but when the media is packed too tightly into the cartridge, only part of the surface area is usable – resulting in both flow restrictions and limited contaminant holding capacity. The 'blind' or unusable area commonly occurs near the inside diameter, where the pleats are packed most tightly. The Betafine™ XL Filter Cartridge features our Advanced Pleat Technology (APT) design for extended service life. This design maximizes the useful surface area while maintaining open flow paths between media pleats.
Typical Applications: Machine tool lubrication, detergents, process and waste water, Reverse osmosis (RO) protection and pre–filtration, Solvent, bulk chemical, and process filtration.
The filter media is constructed from continuous micro–fibers that provide a uniform matrix. There are nine distinct absolute–rated filter grades, beginning at 70 µm, ranging to the very fine 0.2 µm rating. Betafine XL filters are absolute–rated to Beta 1000 (99.9% efficiency at each respective rating).