3M™ Zeta Plus™ 40Q Series Filter Cartridge
Supplier: 3M Healthcare
7010381503 7010411036 7010381022
76028-927CS 2888.78 USD
76028-927 76031-465 76031-865
3M™ Zeta Plus™ 40Q Series Filter Cartridge
Filters Filter Cartridges
Zeta Plus™ 40Q Series Purifier Cartridges filter metallic ion contaminants from high purity chemicals such as resins, photoresists, and solvents in a single pass. The purifier contains ion exchange groups bonded within a porous depth media structure, enhancing surface area and providing metallic contaminant reduction to single digit ppb levels.
  • Filters metallic ion contaminants from high purity chemicals, such as resins, photoresists, and solvents
  • Reduces trace metals to single-digit ppb levels
  • Graded density construction increases contact time between the fluid and reactive sites of the purifier cartridge
  • High flow rates provide increased throughputs as compared to ion exchange columns
  • Superior metals removal capacity
Zeta Plus™ 40Q series purifiers are designed in a wide range of configurations, allowing the same filter media to be used throughout laboratory, pilot, and full production scale-up.
Zeta Plus™ 40Q series purifier cartridges vs ion exchange columns.
Ion exchange columns, a process step commonly used to remove trace metal ions from high purity chemicals, are often the limiting factor in reducing the manufacturing cycle. Even with low flow rates and long residence times, ion exchange columns typically cannot reduce the trace metal content to the required specification levels in a single pass. One reason for this is that the ion exchange resin beads have a relatively low surface area to volume ratio, and another reason is that a large void volume exists between each resin bead. However, we have developed a chemically-modified purifier, capable of filtering metallic or ionic contaminants from photoresists, resins, solvents, and ancillary chemicals, with favorable flow rates and throughput. We accomplish this by maximizing purification surface area, which reduces the potential for channeling and increases the contact time between the chemical and reactive sites of the purifier cartridge.