3M™ High Flow Series Replacement Cartridge, Model HF20-S, 5615103
Supplier: 3M Healthcare
3M™
7000125416
76030-327CS 1514.5 USD
76030-327
3M™ High Flow Series Replacement Cartridge, Model HF20-S, 5615103
Filters Filter Cartridges
3M™ Water Filtration Products High Flow Series Model HF20–S Replacement Cartridge provides you with Recipe Quality Water™ for consistent quality ice that can help make any beverage look more appealing. Our replacement cartridge also helps to protect your equipment from the potentially harmful build–up of scale, which can decrease production and profitability.
  • Provides consistent quality water for commercial ice machines by reducing sediment*, chlorine taste and odor, cyst** and scale*
  • Reduces the ability of calcium and magnesium to form as hard scale* on evaporator plates which can reduce downtime and unscheduled service maintenance
  • Reduction of sediment* helps prevent equipment wear and can help extend the life of parts and fittings
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs
Our 3M™ Water Filtration Products High Flow Series Model HF20–S Replacement Cartridge is designed to be effective at reducing sediment*, chlorine taste and odor, cyst** and scale* allowing you to feel more confident in the quality of the products you serve.
The Sanitary Quick Change (SQC) design allows for fast and easy cartridge change–outs that are sanitary by design – requiring no contact with the filter media during cartridge change–out.
3M™ Water Filtration Products branded replacement cartridges and filter systems offer a wide range of solutions designed to address many types of water quality concerns. Third–party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust and quality around the world.
Having cleaner and clearer water for your business is both a pleasure and a reassurance that you're giving your customers recipe quality water. With a water filtration system from 3M Purification, you can provide filtered water that may help protect your equipment and your profits. From commercial point–of–entry to dedicated point–of–use water filtration systems, look to 3M Purification to help you find the right solution.
Ordering information: **As tested with E. Coli ATCC (11229). Tested and verified by manufacturer’s laboratory.