3M™ Micro-Klean™ RT Series Filter Cartridge
Supplier: 3M Healthcare
3M™ Micro-Klean™
7100143559 7100111258 7000125487 7010338060 7000125681 7000029275 7000050952 7010381950 7100066572 7000029276 7000001708 7000125492 7000125488 7010314331 7010381015 7000125485 7000125489 7000125621 7000125477 7000125617 7000029284 7100094723 7000050538 7100011369 7000125476 7010382129 7000148450 7000125491 7010379478 7000125480 7000125696 7000029277 7000125483 7010339545 7100183700 7010339506 7000148449 7000029431 7000050533 7100169693 7100031205 7000050262 7000050537 7000125486 7000029281 7100106902 7100104076 7000125654 RT09Q16G20NN 7000148452 7100038284 7000148467 7000029283 7000125484 7000029417 7010380791 7100031206 7000125490 7010380858 7000148451 7000051046 7000125482 7000125478 7000125658 7100019939 7000029279 7010313968 7010379473 7010315106 7100031207 7010339923 7000050532 7000125481
76034-299CS 1063.51 USD
76034-299 76034-255 76034-331 76034-375 76034-253 76034-373 76034-251 76034-295 76032-803 76034-371 76034-293 76032-801 76034-337 76034-259 76034-335 76034-257 76034-377 76034-291 76034-387 76034-265 76032-799 76034-385 76034-341 76034-263 76032-797 76034-383 76034-381 76034-305 76034-347 76034-303 76034-269 76034-345 76034-267 76032-827 76034-277 76034-397 76034-353 76034-275 76034-395 76034-351 76034-273 76034-393 76032-825 76034-271 76034-391 76032-823 76034-315 76034-357 76034-313 76034-311 76034-355 76034-309 76032-819 76032-817 76032-811 76034-365 76034-243 76034-287 76034-242 76034-285 76034-361 76032-815 76034-283 76032-813 76034-329 76034-327 76034-369 76034-325 76034-367 76034-245 76034-319 76032-807 76032-809
3M™ Micro-Klean™ RT Series Filter Cartridge
Filters Filter Cartridges
Micro-Klean™ RT series filters are all-polypropylene depth filter cartridges offering consistent particle reduction efficiencies, increased surface life, low initial pressure drop and high particle reduction efficiencies. The filter's extended service life results in fewer filter change-outs.
  • Rigid depth filter construction reduces unloading at high differential pressure
  • Enhanced contaminant holding capacity for fewer filter change outs
  • Grooved cartridge with extended surface area promotes fuller utilization of the depth-matrix and promotes long filter life
  • Made without adhesives, binders, surfactants or lubricants for compatibility in a wide range of applications and operating conditions
  • Materials comply with regulations for food and beverage contact and for use in potable water applications
  • Core-less filter structure for ease of disposal via incineration or shredding
  • Continuous integral length filter element
Advancing Depth Filtration through Technological Innovation
The filter's rigid construction provides consistent performance, extended service life and fewer filter change-outs. We manufacture the Micro-Klean™ RT series filters using our own rigid extrusion bonded technology. Our unique manufacturing process combines the superior process control with the quality assurance enabled by an to provide reliable product performance. This exclusive manufacturing process is an ISO 9001 certified quality system and provides a high degree of fiber-to-fiber thermal bonding, without the use of binders, to produce a rigid, core-less filter structure with the following properties:
Does not unload contaminants with increasing differential pressure like typical meltblown filters, Allows grooves to be machined into the upstream surface, without tearing or melting the filter, structure, more than doubling the effective surface area, Exhibits exceptionally low differential pressure for a given filter ratingCore-less filter structure allows for ease of disposal via incineration or shredding, Continuous integral length filter element is easy to install and has no bond joints to break.
Problem
Solution
Our robust Micro-Klean RT series filter captures and retains contaminant within its rigid filter matrix, even under increasing differential pressure. In addition, the unique depth filter structure of the Micro-Klean RT series filter provides a significant increase in contaminant holding capacity and provides greater flow capacity at a given pressure. Unlike soft meltblown and string-wound filters that require core support, our Micro-Klean RT series filter is self-supporting and is grooved to provide greater than twice the surface area. The increase in surface area prevents premature blinding of the outer surface by large particles and gels and promotes fuller utilization of the depth-matrix. The result is significantly longer life than competitive cartridges.
Result
Micro-Kleen RT Series Filter's extended service life results in fewer filter change-outs while its enhanced flow characteristics can typically reduce the number of filters required to achieve a given flow rate. These combined features can significantly reduce total filtration costs.