3M™ AL Series Filter Housing
Supplier: 3M Healthcare
7010338619 7000125561 7000125596 7100054435
76031-808CS 6061.02 USD
76031-808 76031-810 76034-548 76034-550
3M™ AL Series Filter Housing
Filters Filter Cartridges
3M™ AL Series Filter Housing is constructed of durable, cost–effective cast iron and steel to create a quality–built vessel on the outside, designed to satisfy a wide variety of general–purpose industrial and commercial filtration needs. On the inside, long–lasting stainless steel internal components resist corrosion and help make cleaning and maintenance quick and easy.
  • Durable cast iron and steel with stainless steel internals
  • Housings will accept modular 9³/₄" and 10" length cartridges
  • Smart, 3–piece construction disassembles for quick and easy cleaning
More Product Information