3M™ 740 Series Filter Cartridge
Supplier: 3M Healthcare
7100018322 7100018066 7100018323 7100018183 7100018184 7100017829 7100018248 7100018249 7100018339 7000144866 7100018424 7100018320 7100018854 7100018550 7100017739 7100030026 7100041868 7100042909 7100018177 7100018178
76029-206CS 679.05 USD
76029-206 76029-208 76029-210 76029-212 76029-214 76029-216 76029-218 76029-220 76029-222 76029-224 76029-226 76029-228 76029-230 76029-232 76029-234 76029-236 76031-924 76031-926 76031-928 76031-930
3M™ 740 Series Filter Cartridge
Filters Filter Cartridges
3M™ 740 Series filter cartridges are used in applications with high contaminant reduction requirements. Our high efficiency, outside–to–inside flow direction with large diameter, pleated media permits high flow rates.
  • Compound Radial Pleat Design offers high contaminant capacity
  • Microfiber Filtration Media offers high particle reduction efficiency throughout the service life of the filter
  • Higher Flow Capability per Cartridge
  • Consistent and predictable filtration performance
  • No special tools or hardware required for filter change-out
740 series cartridges are used in filter housings that accept 6.5"/165 mm outside diameter and 1.5"/38.1 mm inside diameter filter cartridges with 226 O–ring connections.
740 Series cartridges are offered in two configurations – 'B' and 'K'. The 'B' versions contain the maximum amount of filter media surface area for those applications where high contaminant holding capacity is needed. The 'K' versions contain an optimized amount of filter media surface area (60 to 70% of 'B' versions) for those applications that benefit from a low unit cost filter design. In 'B' and 'K' versions, the microfiber media is pleated in a compound radial design that optimizes the usable surface area of the filter cartridges. The 740 series cartridges also incorporate polypropylene end caps, outer sleeves and cores to protect the pleat structure and provide durability. A double O–ring seal helps reduce the risk of fluid bypass.
Applications: Process Waters (pre–RO, Cooling), Machine Coolants, Chemicals, Refining (Amine, Final Product), Groundwater, Reclaimed Water, Waste Waters, Well Injection and Produced Waters.
PP microfiber forms the basis of the filtration media we use in the 740 series filter cartridges. Our manufacturing processes allow for tightly controlled specifications resulting in a filter media with consistent and predictable particle retention characteristics. We offer 740 series cartridges in micron ratings ranging from 1 to 70 µm (at 99.9% Particle Removal Efficiency). PP provides broad chemical compatibility for use in a variety of applications.
Certifications: Compatible in applications requiring direct food contact in food and beverage processing per 21 CFR.