3M™ Under Sink Drinking Water System
Supplier: 3M Healthcare
7100049871 7100163098
76030-321CS 947.58 USD
76030-321 76030-689
3M™ Under Sink Drinking Water System
Filters Filter Cartridges
The 3M™ Under Sink Drinking Water System, FM1500-B helps reduce asbestos, lead, cysts, chlorine taste and odor, common water-borne bacteria* algae, mold spores** and sediment as a point-of-use filter.
  • Compact design saves space
  • Sanitary quick change (SQC) system eliminates the need to handle filter media
  • FDA CFR-21 compliant materials
  • Does not require electricity to operate
Reduces bacteria through mechanical filtration as opposed to irradiation (as in case of UV) or disinfection methods (as in case of ozone).
*As tested with E.Coli ATCC (11229) [As tested by CUNO Incorporated. Not tested or certifiable by NSF International.]
**As tested with Dekkera bruxellensis ATCC (64276) [As tested by CUNO Incorporated. Not tested or certifiable by NSF International.]