3M™ Zeta Plus™ H Series Filter Cartridge
Supplier: 3M Healthcare
7010338490 7100143017 7000050599 7000050600 7010339345 7000125726 7010379618 7010381555 7010339489 7010314608 7010380765 7000148456 7000125362 7010379621 7000125517 7010379275 7000125518 7100111524
76033-530CS 962.93 USD
76033-530 76033-536 76033-544 76033-548 76028-929 76028-931 76028-939 76028-951 76028-967 76028-969 76028-971 76028-975 76028-977 76028-979 76028-981 76028-983 76028-985 76031-857
3M™ Zeta Plus™ H Series Filter Cartridge
Filters Filter Cartridges
Zeta Plus™ H series filter cartridges utilize a charge-modified depth filter media with high-tensile strength, which performs efficiently under elevated operating temperatures or repeated hot water sanitation cycles. In addition to the mechanical exclusion of particles via depth loading, the filter media adsorbs contaminants too small for mechanical straining alone.
  • High wet-tensile strength filter media withstands multiple steaming and hot water sanitization cycles
  • Extended filter life provides high throughputs, fewer change-outs and reduced operating costs over conventional plate and frame filters
  • Depth filtration and electrokinetic adsorption provide efficient haze and particle reduction at micron ratings smaller than the mechanical rating alone
  • Variety of cartridge sizes offer flexible options for flow requirements
  • All Components are FDA CFR 21 listed for use with food and beverage filter applications
We manufacture Zeta Plus™ H Series Filter Cartridges in 8, 12, and 16 inch diameter, providing filtration surface areas from 2.8 ft² to 34.7 ft² per cartridge.
With a combination of depth filtration and electro-kinetic adsorption, our cartridge provides efficient haze and particle reduction at micron ratings lower than the mechanical rating alone.
A single Zeta Plus H series filter cartridge is composed of an assembly of Zeta Plus filter cells. Each cell is composed of two Zeta Plus H Series filter media discs on either side of a PP 'stiff cell' separator. The discs are sealed together at the circumference by an injection-molded polypropylene edge seal to form a cell. We then unitize the cells into a cartridge, using set compression that results in a rugged, durable construction. The cells are held in place by three stainless steel bands in the core of the cartridge. We manufacture Zeta Plus cartridges in a variety of sizes and number of cell combinations.
Zeta Plus™ filter cartridge system vs. plate and frame filter economics
Plate and frame filters are labor-intensive, requiring two people four to eight hours to change out the media. Zeta Plus filter cartridges are easy to install and remove, usually taking about 15 minutes, netting you significantly less labor cost and allowing you to focus on the work, not the equipment. The savings become substantial when considering the floor space reduction of the vertical Zeta Plus housing design and a typical 50% or greater decrease in initial capital cost when compared to a comparable plate and frame filter.
Zeta Plus H series filter cartridges demonstrate important benefits in terms of effectiveness and operational cost savings, effective particle reduction, Available in a range of media grades, superior strength and resistance, high performance in microbial reduction and low extractables.
Applications
The Zeta Plus H series filter cartridge is ideally suited for clarification and pre-filtration in food and beverage, cosmetic, and general applications where its exceptional high wet-tensile strength media provides extended service life. These include aqueous, alcoholic, proteinaceous, and acidic solutions.