3M™ High Flow Series Filter Cartridge
Supplier: 3M Healthcare
7000051229 7010296495 7000144847 7010382315 7010338749 7000144848 7010382356 7010339932 7000144849 7000125863 7010413523 7010314308 7000051100 7010314295 7010338751 7100036793 7000051027 7100033628 7100001105 7000125690 7000001784 7000051030 7100001103 7000125691 7000051031 7010338448 7100001114 7000001785 7000051032 7010381722 7000125692 7000051033 7100179960 7010381724 7010314111 7000051040 7000051041 7100010510 7010301315 7000051042 7000144846 7010381732 7010409882 7000125695 7100050158 7010338456 7100062948 7000051043 7100106943 7100106933 7100106939 7100044588 7100106931 7100107007 7100106940 7000051525 7100107078 7100106932 7100106941 7000051661 7100002332 7000125935 7010338750 7010303306 7000001802 7010409921 7010303311 7100036258 7010380833 7100001073 7000029433 7100024295 7000125689 7100001072 7000051028 7100009390 7010339564 7000001779 7000029434 7000001780 7010339565 7000020953 7100011209 7000001781 7010415298 7000001782 7000029436 7000001783 7010409154 7010415299 7000029437 7000051034 7000051035 7100003449 7100010544 7000051037 7000125693 7100001841 7000051038 7000051039 7000125694
76032-304CS 226.08 USD
76032-304 76032-306 76032-308 76032-310 76032-312 76032-314 76032-316 76032-318 76032-320 76032-322 76032-324 76032-326 76032-328 76032-330 76032-332 76032-334 76032-336 76032-338 76032-340 76032-342 76032-344 76032-346 76032-348 76032-350 76032-352 76032-354 76032-356 76032-358 76032-360 76032-362 76032-364 76032-366 76032-368 76032-370 76032-372 76032-374 76032-376 76032-378 76032-380 76032-382 76032-384 76032-386 76032-388 76032-390 76032-392 76032-394 76032-396 76032-398 76032-400 76032-402 76032-404 76032-406 76032-408 76032-410 76032-412 76032-414 76032-416 76032-418 76032-420 76032-422 76032-424 76032-426 76033-428 76033-430 76033-432 76033-434 76033-436 76033-438 76033-440 76033-444 76033-446 76033-448 76033-450 76033-452 76033-454 76033-456 76033-458 76033-460 76033-462 76033-464 76033-466 76033-468 76033-470 76033-472 76033-474 76033-698 76033-700 76033-702 76033-704 76033-706 76033-708 76033-710 76033-712 76033-714 76033-716 76033-718 76033-720 76033-722 76033-724 76033-726 76033-728
3M™ High Flow Series Filter Cartridge
Filters Filter Cartridges
3M™ High Flow Series Filter Housings to deliver all the filtration benefits in a compact footprint. Housings are available in standard designs, as well as customizable configurations to suit your specific needs. All standard 3M High Flow Series Filter Housings are designed, manufactured, tested, and code stamped in accordance with ASME section VIII, division 1.
  • 33 to 50% smaller housing size provides better appearance and fit than competitively sized and flow rated housings
  • Robust cartridge centre–post design eliminates bulky support plates providing easy access to housing internals
  • Convenient cover designs provide easy element change–outs
  • Manufactured from carbon steel, 304 or 316L stainless steel offers effective corrosion protection
  • ASME Code design meets high quality and performance standards
3M high flow series filter housings are 33 to 50% smaller than competitively sized housings for a given flow rate.
Our housings are designed with features that make them easy to use and streamline processes. Spend less time servicing thanks to removable cartridge posts that lift out for easy cleaning, which helps maintain cleaner systems and longer product life. Shouldered seal plates fit solidly into the cartridge core for a positive cartridge seal to help prevent bypass, leaks and to maintain system pressure. Housing drains for both sludge and clean chambers feature drain couplings for easy draining. An air vent facilitates effective venting before starting up or draining.
Fewer filters and user–friendly housing designs mean faster, easier change–outs than competitively sized systems. Our high flow series filter housings are available in a variety of sizes to accommodate up to 7 filter elements in both 40–inch and 60–inch lengths. Our horizontal design option maximizes ease of operation and our vertical design can minimize the system's footprint.