3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 40 Gal. Drawdown Tank, 5591203
Supplier: 3M Healthcare
4010046342
76030-234EA 2568.05 USD
76030-234
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 40 Gal. Drawdown Tank, 5591203
Tanks Cylindrical Tanks
3M™ Commercial water storage tank for use with 3M™ reverse osmosis systems, model 40 gal. Drawdown Tank is appropriately sized for boiler-based steamer units up to 300000 BTUs.
  • Reverse osmosis 40 gallon drawdown tank at 60 psi with 15 psi pre-charge
  • Includes 1¼" ball valve assembly
  • Polyethylene tank liner and fiberglass wrap tank exterior
Tanks can be plumbed together to accommodate larger steamer operations. Fittings are designed to accept plumbing from 3M™ reverse osmosis systems.