Wolverine Grinding Wheels, Weiler®
Supplier: Safety & Industrial Supplies
804-56449 804-56455 804-56473 804-56467 804-56468 804-56474 804-56466
300053-392EA 8.52 USD
300053-392 300085-814 300085-820 300053-396 300053-398 300053-402 300085-818
Wolverine Grinding Wheels, Weiler®
Abrasives
  • Designed for general purpose grinding on steel, iron and other ferrous metals
  • Effective for aggressive stock removal
  • Reinforced with fiberglass webbing for added stability