Wolverine Thin Cutting Wheels, Weiler
Supplier: Weiler
804-56067 804-56283 804-56085 804-56083 804-56088 804-56391 804-56013 804-56076 804-56062 804-56015
300053-374EA 3.84 USD
300053-374 300085-738 300085-752 300085-740 300085-754 300085-732 300053-378 300085-734 300085-758 300085-746
Wolverine Thin Cutting Wheels, Weiler
Abrasives
  • Resin-bonded disc is formulated for aggressive stock removal and fast cutting
  • Suitable for general purpose cutting and grinding applications on steel, iron and other ferrous metals
  • Reinforced with fiberglass webbing for stability