Series 182 Flexible Stainless Steel Rulers, Mitutoyo
Supplier: Mitutoyo
504-182-207
300049-462EA 33.23 USD
300049-462
Series 182 Flexible Stainless Steel Rulers, Mitutoyo
Rulers
Easy to read, clear graduations on a satin-chrome finish.
  • Inch/metric measurements
  • Accuracy: ± 0.004/0.0035, ± 0.1/0.09 mm
  • Tempered stainless steel
  • Vinyl sleeve case cluded