RM Single Stage Manifold Regulators, Western Enterprises
Supplier: Safety & Industrial Supplies
312-RM-7-4 312-RM-4-4
300066-952EA 626.74 USD
300066-952 300066-950
RM Single Stage Manifold Regulators, Western Enterprises
Welding Supplies
Pressure compensated single stage design delivers stable delivery pressure performance.
Unique cartridge permits easy, single-unit replacement of vital parts in minutes. This includes easy replacement of the seat, nozzle, filter, spring, seals, and built-in check-relief valve. Cartridge can easily be removed and installed.