Medium Duty Acetylene Tag-A-Long Outfits, Smith Equipment, ORS Nasco
Supplier: ORS Nasco
635-TL-550
300093-048EA 1232.06 USD
300093-048
Medium Duty Acetylene Tag-A-Long Outfits, Smith Equipment, ORS Nasco
Welding Supplies
  • Use with 30-100-540 Oxygen; 30-15-200 Fuel
  • Uses welding nozzle MW205
  • Used on torch handle CW5A
  • 1/4" cutting capacity
  • 1/8" welding range