Proheat™ 1600 STC™ Heat Gun, Master Appliance
Supplier: Master Appliance
Proheat®
467-PH-1600
76006-446EA 1158.07 USD
76006-446
Proheat™ 1600 STC™ Heat Gun, Master Appliance
Heaters Hot Air Dryers
Once a target temperature is reached, the Proheat™ STC™ will automatically regulate the heater to maintain the target temperature
  • Average air velocity: 734 – 4000 FPM
  • Speed up the heating process - work the upper limits of the cure zone
  • Built-in surface temperature sensor and LCD display provide real time display of both target and surface temperatures
  • LTS™ (Laser Targeting System) provides accurate field of view and operating range
  • Dial-in variable temperature and airflow controls
  • Proloc™ supervisor locking feature prevents changing of temperature or airflow for repeatability
  • LED progress indicators indicate heating process
Unit includes a 6', 3-wire grounded, oil resistant neoprene cord.
The unit compensates for changes in the distance from the work surface by automatically adjusting the temperature output.