Proto® Screw Extractors, ORS Nasco
Supplier: Safety & Industrial Supplies
577-9524 577-9526 577-9523 577-9525 577-9527 577-9529 577-9521 577-9525B 577-9528
300003-212EA 21.5 USD
300003-212 300003-218 300003-210 300003-214 300003-220 300003-224 300003-208 300003-216 300003-222
Proto® Screw Extractors, ORS Nasco
Screwdrivers
Proto® Screw extractors with size stamped on extractor refers to drill bit size to be used with that extractor.
  • Drive into the end of broken pipes or fittings
  • Use tap holder or wrench on extractor to turn out the broken part
Order Now
SPECIFICATIONS
Environmentally Preferable