Flashback Arrestor Sets, Western Enterprises

Supplier: Safety & Industrial Supplies
312-FA-10 312-FA-30
300000-057EA 97.58 USD
300000-057 300000-058
Flashback Arrestor Sets, Western Enterprises
Welding Supplies
Internal check valves designed to prevent reverse flow and accidental mixing of oxygen and fuel gases.

  • Color-coded for gas service to provide added safety
  • For use with oxygen/fuel gas lines
  • Torch and regulator models provide downstream and upstream protection
  • 10" maximum cutting capacity

For stopping flashbacks by preventing ignition of mixed gases and to protect equipment such as hoses, regulators, and cylinders.

Certifications: UL Listed.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR