Human Gingival Fibroblasts, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
FC-0095
76035-950EA 951.54 USD
76035-950
Human Gingival Fibroblasts, Lifeline Cell Technology
Cells
Human Gingival Fibroblasts provide an ideal cell system to establish serum free human feeder layers for human embryonic stem cell cultures or as a model to study wound healing, toxicology or basic cell biology.
Isolated from human gingival tissue
Unexposed to antimicrobials or phenol red
Quality tested tor growth and morphology over at least 15 population doublings
Lifeline® Gingival Fibroblasts are isolated from human gingival tissue.
Fibroblast cells have been isolated, plated and expanded in culture vessels twice before being harvested for cryopreservation.
Lifeline® Gingival Fibroblasts are unexposed to antimicrobials or phenol red.
Fibroblasts are quality tested in FibroLife® S2 or FibroLife® Xeno-Free Medium to ensure proper growth and morphology over a period of at least 15 population doublings.
Gingival Fibroblasts can be grown in Xeno-free conditions when cultured in FibroLife® Xeno-Free Medium.