Quick Connect Components, Western Enterprises
Supplier: Safety & Industrial Supplies
312-QDB100 312-QDB201 312-QDB301 312-QDB302 312-QDB303 312-QDB101 312-QDB105 312-QDB300 312-QDB205 312-QDB200 312-QDB106 312-QDB206
300052-408EA 30.83 USD
300052-408 300052-414 300078-808 300078-810 300078-812 300052-410 300078-798 300078-806 300078-802 300052-412 300078-800 300078-804
Quick Connect Components, Western Enterprises
Welding Supplies
  • Brass construction provides long service life
  • Cycle tested over 100,000 operations indicting a service life of 10 to 12 years of comparable use
  • Non-interchangeable oxygen and fuel gas connectors prevent accidental gas mixing
  • Instant closure on reverse flow check values with total shut-off at only 10 oz of back pressure