Sacrifice Perfusion System, Leica
Supplier: Leica
39471001
10030-380EA 4393.91 USD
10030-380
Sacrifice Perfusion System, Leica
Tissue Processors
The Perfusion two automated pressure perfusion system includes an automated air compressor that quietly pumps the air tank of the perfusion one up to 300 mm Hg and stops.
  • Quicker fixation
  • Better tissue quality
  • Better HRP or fluorescence results
  • Avoid soft tissue shrinkage
As each animal is being prepared, the pressure is set up to begin at the turn of a switch. Pressure above physiological pressure is required to clear all blood vessels and leave no red blood cells. This system gives much greater consistency and more reproducable perfusions.
Delivery information: Supplied wirh 4 one liter bottles for solution, lids, 4 sizes of gavage needle.