Saber Tooth Ceramic Flap Discs, Weiler®
Supplier: Safety & Industrial Supplies
804-50107 804-50103 804-50101 804-50102 804-50106 804-50105
300000-928EA 192.33 USD
300000-928 300000-927 300000-923 300000-925 300000-926 300000-924
Saber Tooth Ceramic Flap Discs, Weiler®
Abrasives
  • Fast cut and long life due to the aggressive self sharpening ceramic grain
  • Specially formulated grinding aid prevents dulling and glazing of grain and allows for "cool cutting" protecting high value parts from heat discoloration