Trimmable Tiger Flap Discs, Weiler
Supplier: Safety & Industrial Supplies
804-50002 804-50003 804-50007 804-50006 804-50008
300046-938EA 15.69 USD
300046-938 300046-940 300046-056 300046-054 300046-058
Trimmable Tiger Flap Discs, Weiler
Abrasives
  • This unique Tiger Disc design enables the operator to trim away the back of the disc, exposing more flap abrasive
  • The backing can be trimmed up to 3/8" from the original diameter, dramatically extending the life of the disc