Brass Cylinder Adaptors, Western Enterprises
Supplier: Safety & Industrial Supplies
312-15 312-16 312-306 312-317 312-318 312-320 312-321 312-324 312-415 312-416 312-51 312-61 312-806 312-808 312-810 312-802 312-858 312-809 312-304 312-316 312-820 312-314 312-319 312-322 312-815 312-828 312-315
300007-797EA 27.79 USD
300007-797 300007-805 300007-830 300007-834 300007-835 300007-838 300007-839 300007-841 300007-850 300007-852 300007-859 300007-869 300007-902 300007-903 300007-904 300066-452 300066-456 300066-468 300066-472 300066-480 300066-322 300066-454 300007-828 300007-833 300066-466 300066-478 300007-832 300007-836 300007-840 300066-458 300066-460 300066-462 300066-470 300087-804 300066-475
Brass Cylinder Adaptors, Western Enterprises
Welding Supplies
Use a brass cylinder adaptor to connect a regulator to a cylinder or other equipment when different CGA threads on the mating parts make a direct connection impossible.
Adaptors should not be used as permanent installations.