RediSep Rf Gold® C18 Chromatography Column, Teledyne ISCO
Supplier: Teledyne
692203334 692203328 692203335 692203336 692203337 692203338 692203339 692203341 692203492
101663-832EA 222.77 USD
101663-832 101663-830 101663-834 101663-836 101663-838 101663-840 101663-842 101663-844 101671-884
RediSep Rf Gold® C18 Chromatography Column, Teledyne ISCO
Chromatography Columns
These RediSep Rf Gold® C18 reversed-phase columns are packed with 20 to 40 µm spherical bonded silica, providing improved separation.
The endcapped C18 chains allow up to 20 separations at pH 10 without destroying the silica.