Microtorch with Refillable Fuel Cell, Master Appliance, ORS Nasco
Supplier: ORS Nasco
467-MT-11
300009-157EA 15.84 USD
300009-157
Microtorch with Refillable Fuel Cell, Master Appliance, ORS Nasco
Soldering Supplies Soldering Irons
Unit dimensions are 4.3" x 2.5" L x 1" W.
  • 2500 °F operating temperature
  • Butane gas
  • Adjustable pinpoint flame
  • 20 minute run time
Meets CPSC requirements.
Certifications: Meets CPSC Requirements.