Staticmaster® 2U500 Ionizing Cartridge, 3", NRD
Supplier: NRD
Staticmaster®
2U500
75794-962EA 314.65 USD
75794-962
Staticmaster® 2U500 Ionizing Cartridge, 3", NRD
Ionizers
The Staticmaster® 2U500 Ionizing Cartridge is a self-powered static eliminator with a small profile, making it an ideal solution for in-balance ionization.
  • Self-powered
  • Small profile
  • Easy mounting
  • Ensures thorough static removal
The 2U500 is inherently balanced to zero volts, which helps ensure thorough static removal for accurate weighs every time. It eliminates the problem of "fly-away" powders and inaccurate measurements in precision weighing applications by utilizing its alpha ionizing technology.