V-Gard Protective Caps and Hats, MSA
Supplier: Safety & Industrial Supplies
454-475368 454-454733 454-466356
300008-996EA 29.02 USD
300008-996 300008-978 300054-006
V-Gard Protective Caps and Hats, MSA
Head Protection Safety Helmets
Convenient caps and hats provide comfortable, lightweight protection
  • Comfortable, lightweight protection
  • Self adjusting crown straps
  • Polyethylene shell provides superior impact protection
  • Sturdy polyethylene shell and suspension system work together as a protection system
Caps and hats are ideally suited for applications in arc flash, asbestos abatement, confined space, food processes, manufacturing, painting, sanding and grinding, steel smelting, working at heights, and welding.
Self adjusting crown straps ensure a comfortable fit. Polyethylene shell provides superior impact protection.
Certifications: Meets or Exceeds the Applicable Requirements for a Type 1 Helmet (top impact) outlined in ANSI Z89.1 2003, Class E and G.