Safety Relief Valves, Brass, Western Enterprises
Supplier: Western Enterprises
312-WMV-4-22 312-WMV-4-235 312-WMV-4-350 312-WMV-4-35 312-WMV-4-100 312-WMV-4-50 312-WMV-4-230 312-WMV-8-250
300067-130EA 40.64 USD
300067-130 300067-140 300067-134 300067-144 300067-132 300067-142 300067-138 300067-136
Safety Relief Valves, Brass, Western Enterprises
Welding Supplies
These safety relief valves are used in cryogenic, pressure vessel, manifold, gaseous or liquid nitrogen, oxygen, air and other demanding applications.
  • Silicone seat provides superior sealing/resealing
  • Heavy duty brass construction resists corrosion