Proto® Standard Service Deep Throat, Full Length, Screw C-Clamps, T-Handle, Stanley® Products
Supplier: Stanley
577-408S 577-410 577-412S 577-403S 577-410S 577-412 577-402 577-403
300025-016EA 196.25 USD
300025-016 300025-006 300024-944 300024-930 300025-008 300024-920 300025-014 300025-010
Proto® Standard Service Deep Throat, Full Length, Screw C-Clamps, T-Handle, Stanley® Products
Clamps General Purpose Clamps
Forcing screws fitted with impact-resistant pads.
  • Turns easily under pressure
  • Sliding T-handle on forcing screw
  • Heavy rolled, Acme thread forcing screw
  • Full length forcing screw
  • Black finish
Certifications: CID A-A-429C.