Proto® Super-Duty Long Drive Pin Punches, ORS Nasco
Supplier: Safety & Industrial Supplies
577-48516L 577-48316L 577-48532L 577-48732L 577-48014L
300025-612EA 24.08 USD
300025-612 300025-606 300025-616 300025-620 300025-578
Proto® Super-Duty Long Drive Pin Punches, ORS Nasco
Tool Sets
Modified parabolic striking end for greater resistance to mushrooming and chipping.
Heat-treated for strength and ductility.
Certifications: ANSI Spec. B209.2.