Brass Hose Adaptors, Western Enterprises
Supplier: Western Enterprises
312-41-B 312-124 312-541 312-24 312-558 312-105 312-154 312-C-42 312-103 312-544 312-125 312-253 312-123 312-106 312-234 312-128 312-556 312-540 312-153 312-104 312-40 312-554 312-108 312-545 312-14 312-557 312-24-0 312-107 312-543 312-549 312-235 312-555 312-539 312-537 312-40-B 312-542 312-245 312-244
300087-800EA 11.64 USD
300087-800 300066-206 300066-308 300066-368 300066-348 300066-304 300066-248 300066-204 300066-386 300066-342 300066-264 300066-220 300066-388 300066-222 300007-821 300007-788 300007-865 300007-789 300007-866 300066-370 300007-863 300066-372 300007-864 300007-820 300007-784 300007-861 300007-862 300007-782 300007-783 300066-338 300066-698 300066-312 300066-314 300066-310 300066-376 300066-382 300066-384 300066-262
Brass Hose Adaptors, Western Enterprises
Welding Supplies
Made from brass to ensure functionality and durability, these hose adapters can accommodate up to 200 psi of pressure.
Threads have been manufactured to the highest standards to ensure a tight and secure fitting.