Proto® TorquePlus™ Metric Sockets, ORS Nasco
Supplier: ORS Nasco
577-47045M 577-4713TM 577-5208M 577-5210M 577-4707TM 577-4711M 577-4710M 577-4710TM 577-5217MH 577-5216MH 577-5206MH 577-5212M 577-4705TM 577-4713M 577-5214MH 577-5224MH 577-4709M 577-5222MH 577-4705M 577-5207M 577-5215MH 577-4706M 577-5213M 577-5213MH 577-4714M 577-4709TM 577-5224M 577-5209MH 577-4706TM 577-5217M 577-4711TM 577-5220MH 577-4712M 577-4714TM 577-47055M 577-5226M 577-4708M 577-5218M 577-5206M 577-4707M 577-5211M 577-5210MH 577-5219MH 577-5212MH 577-5220M 577-5211MH 577-4708TM 577-4715M 577-5218MH 577-5209M 577-5225M 577-5207MH 577-5216M 577-5214M 577-5226MH 577-5223M 577-4712TM 577-5219M 577-5208MH 577-47055TM 577-5215M 577-4704M
300025-456EA 11.32 USD
300025-456 300026-182 300025-414 300025-452 300025-410 300025-294 300025-296 300025-330 300026-138 300026-136 300026-174 300026-054 300026-172 300026-052 300026-256 300026-254 300026-050 300025-326 300025-448 300025-288 300025-360 300026-248 300026-246 300026-048 300026-208 300026-206 300026-086 300026-244 300026-168 300026-124 300026-088 300026-122 300026-166 300025-438 300025-436 300025-316 300025-432 300025-356 300025-312 300026-158 300025-282 300026-236 300026-152 300026-150 300026-156 300026-234 300025-302 300025-346 300026-072 300025-426 300026-070 300025-386 300025-300 300025-340 300025-392 300026-104 300026-064 300026-186 300026-062 300026-184 300026-102 300026-188
Proto® TorquePlus™ Metric Sockets, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in metric.