Proto® TorquePlus™ Deep Sockets, ORS Nasco
Supplier: ORS Nasco
577-4718L 577-5026 577-5016 577-5008 577-5020 577-4712LTM 577-5012H 577-5016H 577-5014H 577-4706LT 577-4708LT 577-5024H 577-5030H 577-4707LT 577-47055LTM 577-4709L 577-4708L 577-5032 577-5022 577-5010H 577-4710LT 577-4707L 577-4709LT 577-5022H 577-4712L 577-4714L 577-4706L 577-4711L 577-5024 577-5010 577-5018 577-4714LT 577-4710LTM 577-5020H 577-5008H 577-5030 577-5028 577-5018H 577-4709LTM 577-4718LT 577-5014 577-4711LTM 577-5028H 577-4713LTM 577-4706LTM 577-4712LT 577-4716L 577-5032H 577-5012 577-4705LTM 577-4708LTM 577-4714LTM 577-4716LT 577-4710L 577-4707LTM 577-4711LT
300025-458EA 15.48 USD
300025-458 300025-932 300025-338 300025-892 300031-004 300025-334 300025-972 300031-006 300025-890 300031-008 300026-018 300025-380 300031-010 300031-012 300026-014 300026-012 300002-940 300025-406 300025-968 300025-408 300025-324 300025-446 300025-320 300025-442 300025-960 300025-880 300026-008 300026-002 300025-928 300025-878 300025-914 300025-954 300025-998 300031-024 300025-310 300025-354 300031-026 300025-350 300025-990 300025-918 300025-306 300025-944 300025-900 300025-468 300025-424 300025-986 300025-420 300031-014 300025-982 300025-466 300031-016 300025-460 300025-384 300031-020 300031-022 300025-904
Proto® TorquePlus™ Deep Sockets, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in inches (and metric on select models).
Certifications: ASME B107.5M.