Semi-skirted 96-well PCR Plates, Phenix Research Products
Supplier: Phenix Research Products
1147B52 1147B60 1147B54 1147B56 1147B55
101100-924PK 73.69 USD
101100-924 101100-756 101100-750 101100-752 101100-754
Semi-skirted 96-well PCR Plates, Phenix Research Products
Microplates PCR Plates
These standard semi-skirted 96-well PCR plates are designed to fit thermal cyclers and ABI sequencers.
  • Four raised corners
  • Slightly raised wells for optimum sealing
  • Free of DNase, RNase and human DNA