Proto® TorquePlus™ Deep Impact Sockets, 1", 1" Drive, ORS Nasco
Supplier: Safety & Industrial Supplies
577-10014L 577-10018L 577-10019L 577-10022L 577-10023L 577-10026L 577-10032L 577-10034L 577-10037L 577-10016L 577-10020L 577-10028L 577-10035L 577-10015L 577-10017L 577-10021L 577-10024L 577-10025L 577-10029L 577-10030L 577-10031L 577-10038L 577-10040L 577-10041L 577-10044L 577-10036L
300023-146EA 67.27 USD
300023-146 300023-168 300023-170 300023-184 300023-190 300023-206 300023-234 300030-746 300030-752 300023-158 300023-176 300023-214 300030-748 300023-152 300023-164 300023-180 300023-194 300023-202 300023-218 300023-224 300023-230 300023-254 300023-258 300023-262 300023-274 300030-750
Proto® TorquePlus™ Deep Impact Sockets, 1", 1" Drive, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Impact Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high-performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in inches.