1-Hydroxybenzotriazole monohydrate, (H₂O 10-14%)

Supplier: Advanced ChemTech

Synonyms: HOBT monohydrate1H-1,2,3-Benzotriazol-1-ol monohydrateHOBT hydrate1-Hydroxybenzotriazole hydrate

RC8201-100 RC8201-1000
101116-616EA 319.85 USD
101116-616 101116-614
1-Hydroxybenzotriazole monohydrate, (H₂O 10-14%)
1-Hydroxybenzotriazole monohydrate
Formula: C₆H₅N₃O·1H₂O
MW: 153.14 g/mol
CAS Number: 80029-43-2

Order Now

Specification Test Results

Water content 10 - 14%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR