Regulator Inlet Nipples, Western Enterprises
Supplier: Western Enterprises
312-CO-4 312-15-4SF 312-66 312-83-CV 312-16-3 312-180-3 312-SS-63 312-69 312-15-8CV 312-83 312-15-3SF 312-603 312-663 312-683 312-14-3 312-SS-15-3 312-603-A 312-CO-3 312-SS-333 312-66-SF 312-SS-603A 312-67 312-15-11 312-87 312-13-3 312-73 312-SS-60-3 312-69-SF 312-680-3SF 312-SS-6347-3 312-347-3 312-63 312-305-3A 312-63-CV 312-15-5 312-29-3 312-6-CO-4 312-15-8 312-300-3 312-15-3 312-16-8 312-614-5 312-15-4 312-76 312-SS-CO-3 312-SS-347-3 312-6347-3 312-CO-4CV 312-SS-83 312-615-3 312-SS-15-3CV 312-283 312-CO-8 312-615-3T 312-SS-14-3
300066-408EA 32.74 USD
300066-408 300066-448 300008-014 300007-807 300066-324 300066-444 300007-808 300066-402 300007-803 300007-804 300066-440 300007-966 300007-845 300007-801 300007-802 300007-887 300007-800 300007-965 300007-885 300007-884 300007-882 300067-004 300066-418 300066-258 300067-012 300067-016 300066-428 300007-906 300007-827 300066-424 300007-867 300066-420 300066-266 300007-824 300066-394 300067-022 300067-026 300066-718 300066-998 300066-316 300067-040 300066-714 300066-996 300066-476 300066-234 300007-879 300066-992 300007-876 300007-799 300007-877 300066-482 300007-795 300066-280 300007-871 300067-034
Regulator Inlet Nipples, Western Enterprises
Welding Supplies
Utilize these brass regulator inlet nipples for various gas regulators with a 1/4 NPT connection.
The inlet nipple provides fast and semi-permanent connection, however not as permanent installations. They are made up of a brass material for maximum durability.