Polyethylene glycol monomethyl ether, M.W. 5000 50% (w/v)
Supplier: Rigaku Reagents
Synonyms:
Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH
1008069
1008068
101443-868EA
170
USD
101443-868
101443-476
Polyethylene glycol monomethyl ether, M.W. 5000 50% (w/v)
Polyethylene glycol monomethyl ether
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.
Formula:
CH₃(OCH₂CH₂)nOH |
MDL Number:
MFCD00084416 CAS Number: 9004-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...