Polyethylene glycol 3,000 50% (w/v)

Supplier: Rigaku Reagents

Synonyms: PEG 3,000

1008056 1008057
101443-468EA 70.62 USD
101443-468 101443-860
Polyethylene glycol 3,000 50% (w/v)
Polyethylene glycol 3,000

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: H(OCH₂CH₂)nOH
MW: 3000
Density: 1.21
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR