Polyethylene glycol 300 ≥100% (v/v)

Supplier: Rigaku Reagents

Synonyms: PEG 300

1008412 1008413
101443-466EA 82.38 USD
101443-466 101443-858
Polyethylene glycol 300 ≥100% (v/v)
Polyethylene glycol 300

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: H(OCH₂CH₂)nOH
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR