Polyethylene glycol 200 ≥100% (v/v)

Supplier: Rigaku Reagents

Synonyms: Ethylene glycol polymer average Mn 200PEG 200

1008410 1008411
101443-850EA 151.69 USD
101443-850 101443-458
Polyethylene glycol 200 ≥100% (v/v)
Polyethylene glycol 200

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: H(OCH₂CH₂)nOH
MW: 200 g/mol
MDL Number: MFCD01779596
CAS Number: 25322-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR